Learn More
DR Instruments Hemostatic Forceps with Black Oxide Coating, 5 in., Straight
SDPSCI_ED
Catalog No. S72157
Change view
Click to view available options
Tip Style:
Precision Point
Catalog No. Tip Style
S72157 Precision Point
1 options
Catalog No. S72157 Supplier DR Instruments Supplier No. 45BO
Add to Cart
Add to Cart
Hemostatic Forceps with Black Oxide Coating are made for dissection purposes. Clamping action helps to hold specimen and free up hands for other tasks.
  • With hemostatic forceps you perform many functions, such as hold and pinch delicate veins and nerves, separate tissues.
  • The use of hemostat is not limited to one type of organism but can be applied to various specimens, from small vertebrates to invertebrates, plants, and other biological materials.
Specifications
Product Type Hemostatic Forceps
Coating Black Oxide
Material Stainless Steel
Style Straight
Tip Style Precision Point
Length (English) 5 in.
Length (Metric) 12.7 cm
Weight (English) 0.6 oz.