Learn More
Sklar Instruments Econo Kelly Hemostatic Forceps, 5-1/2 in., Straight, Serrated, Stainless Steel. BX12.
SDP
Catalog No. 501109755 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Kelly Hemostatic Forceps, 5-1/2 in., Straight, Serrated, Stainless Steel. Econo Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 5-1/2 inches. They are sold in boxes of 12.
Catalog No. 50-110-9755 Supplier Sklar Instruments Supplier No. 21436
Econo Kelly Hemostatic Forceps, 5-1/2 in., Straight, Serrated, Stainless Steel. Econo Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 5-1/2 inches. They are sold in boxes of 12.