Learn More
Med Vet International Rochester-Carmalt Forceps, 6 1/4in., Straight, German Grade, Von Klaüs
Catalog No. 502092265
Encompass_Preferred
This 6 1/4in. Rochester-Carmalt forceps is a German grade surgical instrument. Straight with longitudinal serrations and cross-serrated tips. Rochester-Carmalt forceps are a type of hemostatic clamp that is used to stop blood flow of large vessels or to grip tough tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2265 Supplier Med Vet International Supplier No. VK1163616
This 6 1/4in. Rochester-Carmalt forceps is a German grade surgical instrument. Straight with longitudinal serrations and cross-serrated tips. Rochester-Carmalt forceps are a type of hemostatic clamp that is used to stop blood flow of large vessels or to grip tough tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.