Learn More
Hemosure™ iFOB Test Kits: Home Kit Mailers
SDP
Catalog No. 23800522
Change view
Click to view available options
Quantity:
10 Pack
25 Pack
50 Pack
Type:
Home Kit Mailers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-800-522 25 Pack Home Kit Mailers
23-800-515 10 Pack Home Kit Mailers
23-800-502 50 Pack Home Kit Mailers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-800-522 Supplier Hemosure™ Supplier No. PREPACKCM25
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with Hemosure iFOB Test Kits
If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.
Specifications
CPT Code G0328QW, 82274QW
Clia Complexity Waived
Type Home Kit Mailers
For Use With (Application) Immunological fecal occult Blood test
Includes Mailer, 1 collection tube and return form instructions
Quantity 25 Pack
Test Time 5 min.