Learn More
Leica Biosystems HEMOTOXYLIN GILL III 1GAL
Catalog No. NC0882492
Encompass_Preferred
Three Gill formulations are available: The Gill I formula is generally recommended for cytology. The stronger Gill III is usually preferred for use in histology. GIll II is a medium strength hematoxylin. The Gill formula hematoxylins are used in the progressive method of staining. The Gill formulations do not contain mercury. Available in various quantities.
Catalog No. NC0882492 Supplier Leica Biosystems Supplier No. 3801540
Three Gill formulations are available: The Gill I formula is generally recommended for cytology. The stronger Gill III is usually preferred for use in histology. GIll II is a medium strength hematoxylin. The Gill formula hematoxylins are used in the progressive method of staining. The Gill formulations do not contain mercury. Available in various quantities.