Learn More
EUROTROL EuroTrol HemoTrol Duo Control
Catalog No. NC2053156
Encompass
EuroTrol HemoTrol Duo Control - High Level Hemoglobin solution matrix based on a purified hemolysate optimized for the HemoCue Hb 301 and HemoCue Hb 801 Systems. 30-day open vial room temperature stability (2 - 30 °C). 275-day shelf life when refrigerated (2 - 8 °C).
This item is not returnable. View return policy
Catalog No. NC2053156 Supplier EUROTROL Supplier No. 287.003.002
EuroTrol HemoTrol Duo Control - High Level Hemoglobin solution matrix based on a purified hemolysate optimized for the HemoCue Hb 301 and HemoCue Hb 801 Systems. 30-day open vial room temperature stability (2 - 30 °C). 275-day shelf life when refrigerated (2 - 8 °C).