Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EUROTROL Hemoglobin Quality Control Material, 2 Vials, 1.0 mL each, Normal, 2 - 8 °C refrigerated storage, Opened: 31 days, Unopened: 752 days

Catalog No. NC2053154
Encompass

The Eurotrol HemoTrol Duo Normal Quality Control is an assayed control material designed for in vitro diagnostic quality control, specifically for hemoglobin analysis systems. This quality control material includes a purified hemolysate-based hemoglobin solution optimized for accurate quantitative determinations in professional settings. Key benefits include extended stability and minimal lot-to-lot variations, crucial for consistent performance over time.

  • Product type: Quality control material
  • Analyte: Hemoglobin
  • Control level: Normal
  • Volume per vial: 1.0 mL
  • Vials per box: 2
  • Unopened stability: 752 days at 2 - 8 °C
  • Opened stability: 31 days at 2 - 30 °C
  • Storage (unopened): 2 - 8 °C (Refrigerated)
  • Intended use: In vitro diagnostic quality control
  • Regulatory status: Professional use only
  • Analyte target range: 10.7 - 14.3 g/dL

This item is not returnable. View return policy
Catalog No. NC2053154 Supplier EUROTROL Supplier No. 287.002.002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Eurotrol HemoTrol Duo Normal Quality Control is an assayed control material designed for in vitro diagnostic quality control, specifically for hemoglobin analysis systems. This quality control material includes a purified hemolysate-based hemoglobin solution optimized for accurate quantitative determinations in professional settings. Key benefits include extended stability and minimal lot-to-lot variations, crucial for consistent performance over time.

  • Product type: Quality control material
  • Analyte: Hemoglobin
  • Control level: Normal
  • Volume per vial: 1.0 mL
  • Vials per box: 2
  • Unopened stability: 752 days at 2 - 8 °C
  • Opened stability: 31 days at 2 - 30 °C
  • Storage (unopened): 2 - 8 °C (Refrigerated)
  • Intended use: In vitro diagnostic quality control
  • Regulatory status: Professional use only
  • Analyte target range: 10.7 - 14.3 g/dL

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.