Learn More
Andwin Scientific HEMTLGY CELL CNTR VIAL,1000UNT
SDP
Catalog No. 50820930
Encompass_Preferred
The Hematology cell counter vial is ideal for automated blood cell counters or Coulter counters. The vials feature optically clear parallel sides made from high grade polystyrene and graduations at 10 and 20 mL that allow users to measure samples without unwanted contaminants and confusing background. This product is sold with an easily removable, leak-tight polyethylene snap cap with tab. It is sold in a case of 1000.
Catalog No. 50-820-930 Supplier Andwin Scientific Supplier No. 90401000
The Hematology cell counter vial is ideal for automated blood cell counters or Coulter counters. The vials feature optically clear parallel sides made from high grade polystyrene and graduations at 10 and 20 mL that allow users to measure samples without unwanted contaminants and confusing background. This product is sold with an easily removable, leak-tight polyethylene snap cap with tab. It is sold in a case of 1000.