Learn More
ACROBiosystems ACROBiosystems Hendra virus Fusion glycoprotein (A263T), His Tag
Catalog No. 502107187
Encompass_Preferred
Recombinant Protein;500UG;Hendra virus Fusion glycoprotein (A263T), His Tag (FUN-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Accession # O89342-1 (A263T)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.7 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Fusion Protein Antibody, Human IgG1 (5B3) at 1 μg/mL (100 μL/well) can bind Hendra virus Fusion glycoprotein (A263T), His Tag (Cat. No. FUN-H52H4) with a linear range of 1-2 ng/mL (QC tested).
Catalog No. 50-210-7187 Supplier ACROBiosystems Supplier No. FUNH52H4500UG
Recombinant Protein;500UG;Hendra virus Fusion glycoprotein (A263T), His Tag (FUN-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Accession # O89342-1 (A263T)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.7 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Fusion Protein Antibody, Human IgG1 (5B3) at 1 μg/mL (100 μL/well) can bind Hendra virus Fusion glycoprotein (A263T), His Tag (Cat. No. FUN-H52H4) with a linear range of 1-2 ng/mL (QC tested).