Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heneicosafluorodecyl Iodide 98.0+%, TCI America™
SDP

Catalog No. H08445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. H08445G Supplier TCI America Supplier No. H08445G

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 423-62-1
Molecular Formula C10F21I
Molecular Weight (g/mol) 645.981
MDL Number MFCD00001065
InChI Key UDWBMXSQHOHKOI-UHFFFAOYSA-N
Synonym perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
PubChem CID 67920
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
SMILES C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 67°C
Color Red-White
Boiling Point 102°C
Quantity 5 g
Formula Weight 645.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heneicosafluorodecyl Iodide
EINECSNumber (2)-0090
RTECSNumber HD8710000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.