Learn More
Medchemexpress LLC HY-W089845 500mg Medchemexpress, Heneicosane CAS:629-94-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW089845500MG
Encompass_Preferred
Medchemexpress, HY-W089845 500mg Heneicosane CAS:629-94-7 Heneicosane is an aroma component isolated from Streptomyces philanthi RL-1-178 or Serapias cordigera. Heneicosane is a pheromone and inhibits aflatoxin production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3511