Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heneicosane | 629-94-7 | MFCD00009346 | 296.57 | C21H44 | 5 G
SDP

Catalog No. 5000208722
Encompass_Preferred

Heneicosane is an analytical reference standard (C21H44, MW 296.57) used for research and analytical applications, including studies in insect chemical ecology and investigations of anti-inflammatory and antifungal activity.

  • Analytical standard grade for research and analytical assays.
  • Molecular weight 296.57 and formula C21H44.
  • CAS number 629-94-7 for unambiguous identification.
  • Available pack sizes vary by supplier; manufacturer lists up to 1 g, supplier SKU indicates 5 g (pack-size discrepancy).
  • Store under recommended conditions as specified in the certificate of analysis.

Catalog No. 50-002-08722 Supplier Medchemexpress LLC Supplier No. HYW0898455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heneicosane is an analytical reference standard (C21H44, MW 296.57) used for research and analytical applications, including studies in insect chemical ecology and investigations of anti-inflammatory and antifungal activity.

  • Analytical standard grade for research and analytical assays.
  • Molecular weight 296.57 and formula C21H44.
  • CAS number 629-94-7 for unambiguous identification.
  • Available pack sizes vary by supplier; manufacturer lists up to 1 g, supplier SKU indicates 5 g (pack-size discrepancy).
  • Store under recommended conditions as specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.