Learn More
Medchemexpress LLC Heneicosanoic acid | 2363-71-5 | 1 ML
SDP
Catalog No. 5000232845
Encompass_Preferred
Heneicosanoic acid (C21H42O2; MW 326.56 g/mol) is a long-chain saturated fatty acid and endogenous metabolite supplied as a powder or as prepared solutions for analytical and research use.
  • Long-chain saturated fatty acid (C21H42O2).
  • Supplied as powder and as prepared solutions for convenience.
  • Provided in prepared concentrations such as 1, 5, and 10 mM.
  • Soluble in ethanol and DMSO at listed concentrations.
  • Suitable as an analytical standard and research reagent.
Catalog No. 50-002-32845 Supplier Medchemexpress LLC Supplier No. HY12144710MM 1ML
Heneicosanoic acid (C21H42O2; MW 326.56 g/mol) is a long-chain saturated fatty acid and endogenous metabolite supplied as a powder or as prepared solutions for analytical and research use.
  • Long-chain saturated fatty acid (C21H42O2).
  • Supplied as powder and as prepared solutions for convenience.
  • Provided in prepared concentrations such as 1, 5, and 10 mM.
  • Soluble in ethanol and DMSO at listed concentrations.
  • Suitable as an analytical standard and research reagent.