Learn More
Apexbio Technology LLC Heneicosanoic Acid 2363-71-5 200mg
SDP
Catalog No. 5000089321
Encompass_Preferred
Heneicosanoic Acid (CAS 2363-71-5) is a saturated fatty acid consisting of a 21-carbon chain It is naturally present in various biological sources including plant oils animal fats beeswax and certain microorganisms While the precise molecular targets of heneicosanoic acid are not fully elucidated saturated long-chain fatty acids such as this are studied for their roles in membrane structure lipid metabolism and cellular signaling pathways Heneicosanoic acid is utilized in biomedical research to investigate lipid biosynthesis metabolic profiling and as an analytical standard in lipidomics Its presence in natural sources also makes it relevant for studies on nutrition and metabolic disorders
Catalog No. 50-000-89321 Supplier Apexbio Technology LLC Supplier No. B8629200
Heneicosanoic Acid (CAS 2363-71-5) is a saturated fatty acid consisting of a 21-carbon chain It is naturally present in various biological sources including plant oils animal fats beeswax and certain microorganisms While the precise molecular targets of heneicosanoic acid are not fully elucidated saturated long-chain fatty acids such as this are studied for their roles in membrane structure lipid metabolism and cellular signaling pathways Heneicosanoic acid is utilized in biomedical research to investigate lipid biosynthesis metabolic profiling and as an analytical standard in lipidomics Its presence in natural sources also makes it relevant for studies on nutrition and metabolic disorders