Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heneicosanoic acid | 2363-71-5 | 98.0% | 500 MG
SDP

Catalog No. 5000230551
Encompass_Preferred

Heneicosanoic acid is a long-chain saturated fatty acid supplied as a research-grade analytical standard. It occurs naturally in plants and animals and is used in lipid analysis and metabolomics workflows.

  • Purity 98.0%.
  • Chemical formula C21H42O2; molecular weight 326.56 g/mol.
  • White to off-white solid physical form.
  • Suitable as an analytical standard for lipid profiling and chromatographic reference.
  • Supplied in a 500 MG quantity for laboratory use.

Catalog No. 50-002-30551 Supplier Medchemexpress LLC Supplier No. HY121447500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heneicosanoic acid is a long-chain saturated fatty acid supplied as a research-grade analytical standard. It occurs naturally in plants and animals and is used in lipid analysis and metabolomics workflows.

  • Purity 98.0%.
  • Chemical formula C21H42O2; molecular weight 326.56 g/mol.
  • White to off-white solid physical form.
  • Suitable as an analytical standard for lipid profiling and chromatographic reference.
  • Supplied in a 500 MG quantity for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.