Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heneicosanoic acid, ≥99% (capillary GC), MilliporeSigma™ Supelco™

Catalog No. 111013635 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3635 100 mg
11-101-3634 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3635 Supplier MilliporeSigma Supelco Supplier No. H5149100MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 2363-71-5
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)19 COOH
MDL Number MFCD00002805
Melting Point 74°C to 75°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 326.56
Percent Purity ≥99% (capillary GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.