Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Henicosan-11-ol | 3381-26-8 | MFCD00026547 | 98.0% | 312.57 g/mol | C21H44O | 5 G
SDP

Catalog No. 5000282456
Encompass_Preferred

Henicosan-11-ol (11-Heneicosanol; CAS 3381-26-8) is a long-chain primary alcohol used as a reagent in synthetic chemistry and research. Supplied as a white to off-white solid, it is suitable for esterification and other organic transformations.

  • High purity (98.0%).
  • Molecular formula C21H44O; molecular weight 312.57 g/mol.
  • Appearance: white to off-white solid.
  • Available in small laboratory pack sizes including 5 G.
  • Store at room temperature for long-term stability; in solvent keep at -80°C (2 years) or -20°C (1 year).
  • Commonly used as an alcohol reagent for ester synthesis and lipid research.

Catalog No. 50-002-82456 Supplier Medchemexpress LLC Supplier No. HYW2826635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Henicosan-11-ol (11-Heneicosanol; CAS 3381-26-8) is a long-chain primary alcohol used as a reagent in synthetic chemistry and research. Supplied as a white to off-white solid, it is suitable for esterification and other organic transformations.

  • High purity (98.0%).
  • Molecular formula C21H44O; molecular weight 312.57 g/mol.
  • Appearance: white to off-white solid.
  • Available in small laboratory pack sizes including 5 G.
  • Store at room temperature for long-term stability; in solvent keep at -80°C (2 years) or -20°C (1 year).
  • Commonly used as an alcohol reagent for ester synthesis and lipid research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.