Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Henicosanoic acid | 2363-71-5 | 98.0% | 250 MG
SDP

Catalog No. 5000232083
Encompass_Preferred

Heneicosanoic acid is a long-chain saturated fatty acid supplied as an analytical standard for research and metabolite analysis. It is provided as a solid, white to off-white powder with high purity suitable for chromatographic and mass-spectrometry workflows.

  • Long-chain saturated fatty acid (C21H42O2), CAS 2363-71-5.
  • Molecular weight 326.56 g/mol.
  • Purity 98.0%.
  • Available as 250 MG and additional pack sizes for analytical use.
  • Solid form: white to off-white powder, suitable for preparation of solutions.
  • Storage: powder stability at -20 °C (up to 3 years) and 4 °C (up to 2 years); shortened stability in solvent.
  • Intended for research and analytical applications as an endogenous metabolite or analytical standard.

Catalog No. 50-002-32083 Supplier Medchemexpress LLC Supplier No. HY121447250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heneicosanoic acid is a long-chain saturated fatty acid supplied as an analytical standard for research and metabolite analysis. It is provided as a solid, white to off-white powder with high purity suitable for chromatographic and mass-spectrometry workflows.

  • Long-chain saturated fatty acid (C21H42O2), CAS 2363-71-5.
  • Molecular weight 326.56 g/mol.
  • Purity 98.0%.
  • Available as 250 MG and additional pack sizes for analytical use.
  • Solid form: white to off-white powder, suitable for preparation of solutions.
  • Storage: powder stability at -20 °C (up to 3 years) and 4 °C (up to 2 years); shortened stability in solvent.
  • Intended for research and analytical applications as an endogenous metabolite or analytical standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.