Learn More
Cayman Chemical Heneicosapentaenoic Acid methyl ester, Purity: ≥98%, CAS: 65919-53-1, 10 mg
SDP
Catalog No. 502952632
Encompass_Preferred
A prodrug formulation of HPA which may be useful in various biochemical studies and as a reference standard in analytical work
Catalog No. 50-295-2632 Supplier Cayman Chemical Supplier No. 1162210 MG
A prodrug formulation of HPA which may be useful in various biochemical studies and as a reference standard in analytical work