Learn More
Cayman Chemical Heneicosanoic Acid, Purity: ≥98%, CAS: 2363-71-5, 500 mg
SDP
Catalog No. 502987732
Encompass_Preferred
A very long-chain saturated fatty acid; found in plants and animals, including human milk fat; also found in LPS from Gram-negative bacteria
Catalog No. 50-298-7732 Supplier Cayman Chemical Supplier No. 22593500 MG
A very long-chain saturated fatty acid; found in plants and animals, including human milk fat; also found in LPS from Gram-negative bacteria