Learn More
Cayman Chemical Heneicosanoic Acid methyl ester, Purity: ≥98%, CAS: 6064-90-0, 250 mg
SDP
Catalog No. 502934887
Encompass_Preferred
An ester form of heneicosanoic acid; has been used as an internal standard in the quantification of fatty acids in dairy products and fatty acid methyl esters in fish by GC-MS
Catalog No. 50-293-4887 Supplier Cayman Chemical Supplier No. 26865250 MG
An ester form of heneicosanoic acid; has been used as an internal standard in the quantification of fatty acids in dairy products and fatty acid methyl esters in fish by GC-MS