Learn More
Medchemexpress LLC Hentriacontane | 630-04-6 | 98.0% | 436.84 | 250 MG
SDP
Catalog No. 5000425819
Encompass_Preferred
Hentriacontane is a long-chain alkane that exhibits anti-inflammatory, antitumor, and antibacterial activities. It exerts its anti-inflammatory effects by inhibiting the NF-κB pathway.
  • Exhibits anti-inflammatory, antitumor, and antibacterial activities
  • Decreases TNF-α, IL-6, and IL-1β inflammatory parameters in RAW 264.7 cells
  • Increases phosphorylation of NF-kB p65 in RAW 264.7 cells
  • Inhibits inflammatory cytokines in LPS-induced mouse inflammation models
  • Solid appearance, white to off-white color
  • Store at 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)
Catalog No. 50-004-25819 Supplier Medchemexpress LLC Supplier No. 250MGHYN9971
Hentriacontane is a long-chain alkane that exhibits anti-inflammatory, antitumor, and antibacterial activities. It exerts its anti-inflammatory effects by inhibiting the NF-κB pathway.
  • Exhibits anti-inflammatory, antitumor, and antibacterial activities
  • Decreases TNF-α, IL-6, and IL-1β inflammatory parameters in RAW 264.7 cells
  • Increases phosphorylation of NF-kB p65 in RAW 264.7 cells
  • Inhibits inflammatory cytokines in LPS-induced mouse inflammation models
  • Solid appearance, white to off-white color
  • Store at 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)