Learn More
ATCC Hep 3B2.1-7; Hepatocellular Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HB8064
Encompass
Hep 3B2.1-7 [Hep 3B, Hep-3B, Hep3B] is a cell line exhibiting epithelial morphology that was isolated from liver tissue derived from an 8-year-old, Black youth with liver cancer. This cell line contains an integrated hepatitis B virus genome.
Catalog No. 50-238-4185