Learn More
Enzo Life Sciences Hepatitis C core protein monoclonal antibody (C7-50) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804277C100
Encompass_Preferred
Clone: C7-50. Isotype: IgG1. Immunogen: Purified HCV (hepatitis C virus) core fusion protein (genotype 1b). Species reactivity: Hepatitis C virus. Specificity: Recognizes an epitope between aa 21-40 of HCV core protein. This sequence is conserved among different HCV strains. Applications: ELISA, ICC, IP, WB. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9852