Learn More
ATCC Hep G2; Hepatocellular; Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HB8065
Encompass
Hep G2 [HEPG2] is a cell line exhibiting epithelial-like morphology that was isolated from a hepatocellular carcinoma of a 15-year-old, White, male youth with liver cancer. The cell line was deposited by the Wistar Institute and is a suitable transfection host. Expression markers include insulin; insulin-like growth factor II (IGF II).
Catalog No. 50-238-4862