Learn More
ATCC Hep G2, Frozen
Supplier:  ATCC HB8065/DEL
Encompass
These cells are suitable as a transfection host. The cells express 3-hydroxy-3-methylglutaryl-CoA reductase and hepatic triglyceride lipase activities. The cells demonstrate decreased expression of apoA-I mRNA and increased expression of catalase mRNA in response to gramoxone (oxidative stress). There is no evidence of a Hepatitis B virus genome in this cell line.For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188729NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-729FP