Learn More
Active Motif Hep G2 whole-cell extract
SDP
Catalog No. NC1847593
Encompass
Active Motif's Hep G2 whole-cell extract was prepared from cell cultures of the human hepatocellular liver carcinoma Hep G2 cell line. These cells are epithelial in morphology and can be induced to exhibit polarization under appropriate culture conditions. Hep G2 cells are most commonly used for studies of hepatocyte function. Because of their ability to differentiate into polarized epithelium, these cells are routinely utilized to investigate intracellular protein trafficking, particularly in relation to human liver diseases. Hep G2 is the most commonly used cell line in studies pertaining to the regulation of hepatic protein synthesis, particularly the synthesis of acute phase proteins after the onset of a systemic inflammatory response. In addition, these cells are important for studies related to liver metabolism, liver cancers, liver regeneration, liver cytotoxicity, apoptosis and as a model for hepatitis B virus (HBV) viral etiology.
Catalog No. NC1847593 Supplier Active Motif Supplier No. 36412
Active Motif's Hep G2 whole-cell extract was prepared from cell cultures of the human hepatocellular liver carcinoma Hep G2 cell line. These cells are epithelial in morphology and can be induced to exhibit polarization under appropriate culture conditions. Hep G2 cells are most commonly used for studies of hepatocyte function. Because of their ability to differentiate into polarized epithelium, these cells are routinely utilized to investigate intracellular protein trafficking, particularly in relation to human liver diseases. Hep G2 is the most commonly used cell line in studies pertaining to the regulation of hepatic protein synthesis, particularly the synthesis of acute phase proteins after the onset of a systemic inflammatory response. In addition, these cells are important for studies related to liver metabolism, liver cancers, liver regeneration, liver cytotoxicity, apoptosis and as a model for hepatitis B virus (HBV) viral etiology.