Learn More
AEROCOLLOID LLC HEPA CAPSULE FILTER INLINE 3/8
Supplier:  AEROCOLLOID LLC HC214NPTF
Encompass
HEPA Capsule Filter, Inline Filter, Borosilicate Glass Microfibers, pp, 50 psig at 70°F, 3/8'' NPTF
Catalog No. NC1411994