Learn More
MSC Nilfisk Cleanroom Canister Vacuum, 3.25 Gallon, ULPA Filtration, Stainless Steel, 115V
Catalog No. 502838676
Encompass
The IVT1000CR cleanroom canister vacuum is engineered for critical environments, providing high-quality filtration and quiet operation to maintain air purity. Its robust stainless steel construction ensures durability and allows for autoclavable components for sterilization.
  • Standard ULPA filtration
  • AISI 316 stainless steel construction
  • Autoclavable trolley and container
  • Quiet 61 dB operation
  • Compact and lightweight design
  • Sealed filtration system
  • Modular construction for easy maintenance
  • 30 ft. power cord
  • Compatible with cleanrooms down to ISO 4 (Class 10)
Catalog No. 50-283-8676 Supplier MSC Supplier No. 35210384
The IVT1000CR cleanroom canister vacuum is engineered for critical environments, providing high-quality filtration and quiet operation to maintain air purity. Its robust stainless steel construction ensures durability and allows for autoclavable components for sterilization.
  • Standard ULPA filtration
  • AISI 316 stainless steel construction
  • Autoclavable trolley and container
  • Quiet 61 dB operation
  • Compact and lightweight design
  • Sealed filtration system
  • Modular construction for easy maintenance
  • 30 ft. power cord
  • Compatible with cleanrooms down to ISO 4 (Class 10)