Learn More
USA Scientific Inc 18 inch HEPA Filter for 24 inch and 48 inch AirClean
Supplier:  USA Scientific Inc 42343718
Encompass
18 in replacement HEPA filter for 48 in AirClean (2 required) and 24 in (1 required) combination PCR workstation.
Catalog No. NC1970519