Learn More
MSC Nilfisk Wet/Dry Cleaner, HEPA Filter, 12 gal Capacity, Electric
Catalog No. 502836645
Encompass
The Eliminator Pro II wet/dry vacuum is engineered for robust, regular use in challenging environments, delivering consistent performance, powerful suction, and effective filtration. This lightweight and maneuverable single-phase vacuum offers an efficient and economical solution for both wet and dry collection tasks.
  • Designed for rugged applications.
  • 12-gallon collection container with drain hose.
  • Multi-stage filtration, including HEPA and optional dust bag.
  • Efficient bypass motor for minimal maintenance.
  • Quiet operation.
  • Easy to use and service.
  • 50 ft. power cord.
  • Includes 8 ft. hose, stainless steel extension wand, and various nozzles.
Catalog No. 50-283-6645 Supplier MSC Supplier No. 35210350
The Eliminator Pro II wet/dry vacuum is engineered for robust, regular use in challenging environments, delivering consistent performance, powerful suction, and effective filtration. This lightweight and maneuverable single-phase vacuum offers an efficient and economical solution for both wet and dry collection tasks.
  • Designed for rugged applications.
  • 12-gallon collection container with drain hose.
  • Multi-stage filtration, including HEPA and optional dust bag.
  • Efficient bypass motor for minimal maintenance.
  • Quiet operation.
  • Easy to use and service.
  • 50 ft. power cord.
  • Includes 8 ft. hose, stainless steel extension wand, and various nozzles.