Learn More
MSC Nilfisk Industrial Vacuum, Wet/Dry, HEPA Filter, 15 gal, Cleanroom
Catalog No. 502839096
Encompass
The VT60CR Wet/Dry Cleanroom Industrial Vacuum offers contamination control and powerful suction for cleanroom settings. Its four-stage HEPA filtration system handles both wet and dry debris without compromising integrity, making it ideal for pharmaceutical, biotech, and technology manufacturing.
  • HEPA filtration captures 99.97% of microparticles.
  • Main filter protects HEPA from moisture/corrosion.
  • Wet/dry cleaning handles spills without compromising filtration.
  • Prevents clogs with pliable inlet tube and foam filter.
  • Cleanroom accessory kit with wands, hoses, nozzles.
  • Anti-static design minimizes static discharge.
  • Powerful, quiet motor provides robust suction.
  • Detachable trolley with anti-static wheels, easy maneuverability.
  • Simple maintenance: fast filter changes, tank emptying.
Catalog No. 50-283-9096 Supplier MSC Supplier No. 35210442
Add to Cart
Add to Cart
Request a Quote
The VT60CR Wet/Dry Cleanroom Industrial Vacuum offers contamination control and powerful suction for cleanroom settings. Its four-stage HEPA filtration system handles both wet and dry debris without compromising integrity, making it ideal for pharmaceutical, biotech, and technology manufacturing.
  • HEPA filtration captures 99.97% of microparticles.
  • Main filter protects HEPA from moisture/corrosion.
  • Wet/dry cleaning handles spills without compromising filtration.
  • Prevents clogs with pliable inlet tube and foam filter.
  • Cleanroom accessory kit with wands, hoses, nozzles.
  • Anti-static design minimizes static discharge.
  • Powerful, quiet motor provides robust suction.
  • Detachable trolley with anti-static wheels, easy maneuverability.
  • Simple maintenance: fast filter changes, tank emptying.