Learn More
MSC Nilfisk Wet/Dry Vac, HEPA, 5 gal, Stainless Steel, 1 ea
Catalog No. 502866983
Encompass
The Nilfisk AERO series compact wet/dry vacuum cleaner is designed for long-lasting performance, strong suction, low noise levels, and ergonomic operation. This 20-liter machine features a compact design with user-friendly features, ensuring safe operation, easy storage, and increased productivity.
  • Stainless steel collection container
  • Push&Clean system for fast and simple filter cleaning
  • HEPA filtration for safe collection of hazardous materials
  • Washable PET fleece filter with minimum 99.9% filtration efficiency
  • MultiFit accessory system for ultimate flexibility
  • Convenient accessory and cable storage
  • Exhaust air blow function
  • Integrated container handle for easy disposal and lifting
  • Auto start for power tools
  • Stable wheels and ergonomic design for safe operation
  • Fleece bags for easy clean-up of debris (efficiency: 47.3% @ 0.3 microns)
Catalog No. 50-286-6983 Supplier MSC Supplier No. 35210475
The Nilfisk AERO series compact wet/dry vacuum cleaner is designed for long-lasting performance, strong suction, low noise levels, and ergonomic operation. This 20-liter machine features a compact design with user-friendly features, ensuring safe operation, easy storage, and increased productivity.
  • Stainless steel collection container
  • Push&Clean system for fast and simple filter cleaning
  • HEPA filtration for safe collection of hazardous materials
  • Washable PET fleece filter with minimum 99.9% filtration efficiency
  • MultiFit accessory system for ultimate flexibility
  • Convenient accessory and cable storage
  • Exhaust air blow function
  • Integrated container handle for easy disposal and lifting
  • Auto start for power tools
  • Stable wheels and ergonomic design for safe operation
  • Fleece bags for easy clean-up of debris (efficiency: 47.3% @ 0.3 microns)