Learn More
Aviva Systems Biology Corp Hepacam2 Antibody - N-terminal region (ARP44869_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44869_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hepacam2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Rat Hepacam2. Alternate names: RGD1305697ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-165