Learn More
Selleck Chemical LLC Heparan Sulfate
SDP
Catalog No. 5000000777
Encompass_Preferred
Heparan sulfate (HS Heparitin sulfate Alpha-idosane HHS 5 N-Acetylheparan Sulfate Suleparoid Tavidan) a constituent of HS proteoglycans (HSPGs) is a linear polysaccharide present on the cell surface Heparan sulfate influences the binding affinity of intestinal epithelium cells (IECs) to Wnt thereby promoting activation of canonical Wnt signaling and facilitating regeneration of small intestinal crypts after epithelial injury
Catalog No. 50-000-00777 Supplier Selleck Chemical LLC Supplier No. S599225MG
Heparan sulfate (HS Heparitin sulfate Alpha-idosane HHS 5 N-Acetylheparan Sulfate Suleparoid Tavidan) a constituent of HS proteoglycans (HSPGs) is a linear polysaccharide present on the cell surface Heparan sulfate influences the binding affinity of intestinal epithelium cells (IECs) to Wnt thereby promoting activation of canonical Wnt signaling and facilitating regeneration of small intestinal crypts after epithelial injury