Learn More
Selleck Chemical LLC Heparan Sulfate
SDP
Supplier:  Selleck Chemical LLC S59921G
Encompass_Preferred
Heparan sulfate (HS Heparitin sulfate Alpha-idosane HHS 5 N-Acetylheparan Sulfate Suleparoid Tavidan) a constituent of HS proteoglycans (HSPGs) is a linear polysaccharide present on the cell surface Heparan sulfate influences the binding affinity of intestinal epithelium cells (IECs) to Wnt thereby promoting activation of canonical Wnt signaling and facilitating regeneration of small intestinal crypts after epithelial injury
Catalog No. 50-000-04062