Learn More
Amsbio LLC Heparan Sulfate
Catalog No. 502041324
Encompass
Major disaccharide units: GlcA-GlcNAc and IdoA/GlcA-GlcNS (variable O-sulphation). Degree of N-sulphation 65 (N-sulphates/100 disaccharides). Ratio N-sulphation:O-sulphation = 0.73. Single band on agarose gel electrophoresis. Contains both low and high sulphated heparan sulphates.
Catalog No. 50-204-1324 Supplier Amsbio LLC Supplier No. AMS.GAGHS01
Major disaccharide units: GlcA-GlcNAc and IdoA/GlcA-GlcNS (variable O-sulphation). Degree of N-sulphation 65 (N-sulphates/100 disaccharides). Ratio N-sulphation:O-sulphation = 0.73. Single band on agarose gel electrophoresis. Contains both low and high sulphated heparan sulphates.