Learn More
Selleck Chemical LLC Heparan Sulfate 5mg
SDP
Supplier:  Selleck Chemical LLC S59925MG
Encompass_Preferred
Heparan sulfate (HS, Heparitin sulfate, Alpha-idosane, HHS 5, N-Acetylheparan Sulfate, Suleparoid, Tavidan), a constituent of HS proteoglycans (HSPGs), is a linear polysaccharide present on the cell surface. Heparan sulfate influences the binding affinity of intestinal epithelium cells (IECs) toWnt, thereby promoting activation of canonical Wnt signaling and facilitating regeneration of small intestinal crypts after epithelial injury. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3173