Learn More
Medchemexpress LLC Heparan Sulfate-6-O- 100ug | 100ug
SDP
Catalog No. 5000194803
Encompass_Preferred
Heparan Sulfate-6-O-Sulfotransferase-3 (HS6ST3) is an enzyme involved in modifying heparan sulfate proteoglycan biosynthesis It is predicted to be a membrane-bound integral component HS6ST3 shows specific expression in the adult cortex (RPKM 1 3) and frontal lobe (RPKM 0 8) implying its potential role in determining the sulfation patterns of heparan sulfate in these contexts Heparan Sulfate-6-O-Sulfotransferase-3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Heparan Sulfate-6-O-Sulfotransferase-3 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-94803 Supplier Medchemexpress LLC Supplier No. HYP79181100UG
Heparan Sulfate-6-O-Sulfotransferase-3 (HS6ST3) is an enzyme involved in modifying heparan sulfate proteoglycan biosynthesis It is predicted to be a membrane-bound integral component HS6ST3 shows specific expression in the adult cortex (RPKM 1 3) and frontal lobe (RPKM 0 8) implying its potential role in determining the sulfation patterns of heparan sulfate in these contexts Heparan Sulfate-6-O-Sulfotransferase-3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Heparan Sulfate-6-O-Sulfotransferase-3 protein expressed by HEK293 with N-His labeled tag