Learn More
Medchemexpress LLC Heparan sulfate | 9050-30-0 | 593.47 g/mol | C12H19NO20S3 | 500 MG
SDP
Catalog No. 5000245546
Encompass_Preferred
Heparan sulfate is a linear, sulfated glycosaminoglycan used in biochemical and cell biology research to study extracellular matrix interactions, growth factor binding, and cell signaling. The material is supplied as a research-grade polysaccharide (monomer MW 593.47 g/mol) and is water soluble (≈100 mg/mL with sonication). Store dry at 4°C; in solution, store at -80°C for up to 6 months.
  • Linear sulfated glycosaminoglycan suitable for biochemical assays.
  • Interacts with growth factors, morphogens, and adhesion molecules.
  • High aqueous solubility; sonication may be required for dissolution.
  • Multiple package sizes available for small-scale and preparative work.
  • Storage: 4°C dry; in solution -80°C for long-term stability.
Catalog No. 50-002-45546 Supplier Medchemexpress LLC Supplier No. HY101916500MG
Heparan sulfate is a linear, sulfated glycosaminoglycan used in biochemical and cell biology research to study extracellular matrix interactions, growth factor binding, and cell signaling. The material is supplied as a research-grade polysaccharide (monomer MW 593.47 g/mol) and is water soluble (≈100 mg/mL with sonication). Store dry at 4°C; in solution, store at -80°C for up to 6 months.
  • Linear sulfated glycosaminoglycan suitable for biochemical assays.
  • Interacts with growth factors, morphogens, and adhesion molecules.
  • High aqueous solubility; sonication may be required for dissolution.
  • Multiple package sizes available for small-scale and preparative work.
  • Storage: 4°C dry; in solution -80°C for long-term stability.