Learn More
Medchemexpress LLC Heparan sulfate | 9050-30-0 | 593.47 (monomer) | C12H19NO20S3 (monomer) | 1 G
SDP
Catalog No. 5000248755
Encompass_Preferred
Heparan sulfate is a linear, sulfated glycosaminoglycan supplied as a research-grade polysaccharide for investigating cell-surface proteoglycan functions, growth factor interactions, and extracellular matrix biology. Provided as a dry material for laboratory research use only.
  • Linear sulfated glycosaminoglycan derived from animal tissue.
  • Monomer formula C12H19NO20S3 and monomer molecular weight 593.47.
  • Available in common laboratory quantities, including 1 G.
  • Used to study proteoglycan interactions, growth factor binding, and extracellular matrix biology.
  • Supplied for research use only; follow the provided safety and handling guidance.
Catalog No. 50-002-48755 Supplier Medchemexpress LLC Supplier No. HY1019161G
Heparan sulfate is a linear, sulfated glycosaminoglycan supplied as a research-grade polysaccharide for investigating cell-surface proteoglycan functions, growth factor interactions, and extracellular matrix biology. Provided as a dry material for laboratory research use only.
  • Linear sulfated glycosaminoglycan derived from animal tissue.
  • Monomer formula C12H19NO20S3 and monomer molecular weight 593.47.
  • Available in common laboratory quantities, including 1 G.
  • Used to study proteoglycan interactions, growth factor binding, and extracellular matrix biology.
  • Supplied for research use only; follow the provided safety and handling guidance.