Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heparan sulfate | 9050-30-0 | 593.47 (monomer) | C12H19NO20S3 (monomer) | 25 MG
SDP

Catalog No. 5000248147
Encompass_Preferred

Heparan sulfate is a complex, linear glycosaminoglycan used in biochemical and cell biology research to probe growth factor binding, cell signaling, and extracellular matrix interactions. Supplied as a dry material with reported monomer parameters for reference, it dissolves in aqueous buffers for use in assays and cell studies.

  • Supports study of growth factor signaling, including FGFR and Wnt pathways.
  • Suitable for extracellular matrix and proteoglycan research.
  • Soluble in water and PBS; ultrasonic agitation may be required for complete dissolution.
  • Monomer molecular formula C12H19NO20S3 and monomer molecular weight 593.47 for reference.
  • Storage guidance available for sealed material and solutions to preserve stability.

Catalog No. 50-002-48147 Supplier Medchemexpress LLC Supplier No. HY10191625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heparan sulfate is a complex, linear glycosaminoglycan used in biochemical and cell biology research to probe growth factor binding, cell signaling, and extracellular matrix interactions. Supplied as a dry material with reported monomer parameters for reference, it dissolves in aqueous buffers for use in assays and cell studies.

  • Supports study of growth factor signaling, including FGFR and Wnt pathways.
  • Suitable for extracellular matrix and proteoglycan research.
  • Soluble in water and PBS; ultrasonic agitation may be required for complete dissolution.
  • Monomer molecular formula C12H19NO20S3 and monomer molecular weight 593.47 for reference.
  • Storage guidance available for sealed material and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.