Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparanase 1 pre activated
Catalog No. 502905426
Encompass
Recombinant human heparanase-1 is expressed in human HEK 293 cells as an active heterodimer glycoprotein containing a 50 kDa subunit and a 8 kDa subunit. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein. The protein is produced with no artificial tags.
Catalog No. 50-290-5426 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE011610UG
Recombinant human heparanase-1 is expressed in human HEK 293 cells as an active heterodimer glycoprotein containing a 50 kDa subunit and a 8 kDa subunit. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein. The protein is produced with no artificial tags.