Learn More
Lonza Walkersville HepaRG Basal Medium
Catalog No. 501060994
Encompass
HepaRG Basal Medium is a Williams E-based medium without antibiotics for use specifically with the NoSpin HepaRG Cells. Available as 100 mL for direct completion with HepaRG application-dependent supplements.Benefits:Optimized for fully differentiated HepaRG(R) healthApplications:ADMEToxContent:100 mL
Catalog No. 50-106-0994 Supplier Lonza Walkersville Supplier No. MH100
HepaRG Basal Medium is a Williams E-based medium without antibiotics for use specifically with the NoSpin HepaRG Cells. Available as 100 mL for direct completion with HepaRG application-dependent supplements.Benefits:Optimized for fully differentiated HepaRG(R) healthApplications:ADMEToxContent:100 mL