Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ HepaRG™ Induction Medium Supplement (5X)

Catalog No. HPRG740
Encompass_Preferred
Change view
Click to view available options
Quantity:
19 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HPRG740 19 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HPRG740 Supplier Gibco™ Supplier No. HPRG740
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

HepaRG™ Induction Medium Supplement (5X) is a specialized media supplement for use in culturing HepaRG™ cells.

HepaRG™ Induction Medium Supplement (5X) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Induction Medium Supplement (5X) is specifically used for induction of metabolic activity in HepaRG™ cells.

  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

Media Preparation
Working media is prepared by the addition of thawed HepaRG™ Induction Medium Supplement (5X) to 500 mL of William's Medium E (Cat No. A1217601 or 12551032) and 5 mL of GlutaMAX™-I (Cat. No. 35050061). The simple two-step process helps reduce contamination opportunities.

Simplified, Relevant and Reproducible Studies
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Supplement should be stored at -20°C.
Cell Type HepaRG Cells
Concentration 5X
Form Frozen
Product Type Hepatocyte Supplement
Serum Level Standard Serum
Sterility Sterile
Without Additives No Glutamine
For Use With (Application) Drug Metabolism
Purity or Quality Grade Research Grade, Cell Culture Grade
Quantity 19 mL
Shipping Condition Dry Ice
Tested For Sterility Testing (Bacteria and Fungi)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.