Learn More
Lonza Walkersville HepaRG Maintenance and Metabolism Medium Supplement
Catalog No. 501060991
Encompass
HepaRG-NS Maintenance/Metabolism Medium Supplement with corticoids, to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Contains DMSO.Benefits:Ensures high basal metabolic activity in terminally differentiated No Spin HepaRG cellsApplications:ADMEToxContent:14 mL
Catalog No. 50-106-0991 Supplier Lonza Walkersville Supplier No. MHMET
HepaRG-NS Maintenance/Metabolism Medium Supplement with corticoids, to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Contains DMSO.Benefits:Ensures high basal metabolic activity in terminally differentiated No Spin HepaRG cellsApplications:ADMEToxContent:14 mL