Learn More
Lonza Walkersville HepaRG Preinduction and Toxicity Medium Supplement
Catalog No. 501060992
Encompass
HepaRG Pre-induction and Toxicity Medium Supplement with corticoids, to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Completed Pre-induction/Tox Medium is most commonly used for toxicity assays using the NoSpin HepaRG cells.Benefits:Transitional medium for No Spin HepaRG cells just prior to toxicity or induction applicationsApplications:ADMEToxContent:1 conical tube with 12.5mL supplement
Catalog No. 50-106-0992 Supplier Lonza Walkersville Supplier No. MHPIT
HepaRG Pre-induction and Toxicity Medium Supplement with corticoids, to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Completed Pre-induction/Tox Medium is most commonly used for toxicity assays using the NoSpin HepaRG cells.Benefits:Transitional medium for No Spin HepaRG cells just prior to toxicity or induction applicationsApplications:ADMEToxContent:1 conical tube with 12.5mL supplement