Learn More
Lonza Walkersville HepaRG Thawing and Plating Medium Supplement
Catalog No. 501060990
Encompass
HepaRG Thawing and Plating Medium Supplement with corticoids; to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Contains no DMSO.Specifically for use to support thawing and plating of the NoSpin HepaRG Cells.Benefits:Ensures optimal thawing and plating of No Spin HepaRG().Applications:ADMEToxContent:1 tube with 11.8mL supplement
Catalog No. 50-106-0990 Supplier Lonza Walkersville Supplier No. MHTAP
HepaRG Thawing and Plating Medium Supplement with corticoids; to be combined with 100 mL of Basal medium (Catalog #MH100) w/o phenol red. Contains no DMSO.Specifically for use to support thawing and plating of the NoSpin HepaRG Cells.Benefits:Ensures optimal thawing and plating of No Spin HepaRG().Applications:ADMEToxContent:1 tube with 11.8mL supplement