Learn More
Apexbio Technology LLC Heparin(Synonyms: Heparin Sodium, Unfractionated Heparin, Sodium Heparin, Heparinic Acid, Liquaemin, Calciparine), 100mg, CAS: 9005-49-6.
SDP
Catalog No. 5000060625
Encompass_Preferred
Heparin (CAS 9005-49-6) is a highly sulfated glycosaminoglycan known primarily for its anticoagulant properties In biological systems it is stored in granules within specific animal tissues and cells potentially serving defensive roles against pathogens In vitro studies demonstrate that Heparin dose-dependently suppresses granulocyte aggregation and degranulation triggered by FMLP or serum-activated zymosan and specifically blocks FMLP-induced superoxide anion production Additionally in rat models Heparin treatment reduces both the binding affinity and density of angiotensin II receptors on adrenal cells resulting in decreased angiotensin II-stimulated aldosterone secretion Heparin is extensively applied in biomedical research to investigate granulocyte-mediated inflammation and renin-angiotensin system signaling
Catalog No. 50-000-60625 Supplier Apexbio Technology LLC Supplier No. B3602100
Heparin (CAS 9005-49-6) is a highly sulfated glycosaminoglycan known primarily for its anticoagulant properties In biological systems it is stored in granules within specific animal tissues and cells potentially serving defensive roles against pathogens In vitro studies demonstrate that Heparin dose-dependently suppresses granulocyte aggregation and degranulation triggered by FMLP or serum-activated zymosan and specifically blocks FMLP-induced superoxide anion production Additionally in rat models Heparin treatment reduces both the binding affinity and density of angiotensin II receptors on adrenal cells resulting in decreased angiotensin II-stimulated aldosterone secretion Heparin is extensively applied in biomedical research to investigate granulocyte-mediated inflammation and renin-angiotensin system signaling