Learn More
Medchemexpress LLC Heparin | 9005-49-6 | 250 MG
SDP
Catalog No. 5000385313
Encompass_Preferred
Heparin is a highly sulfated glycosaminoglycan that is widely used as an injectable anticoagulant. It possesses the highest negative charge density of any known biological molecule and significantly inhibits exosome-cell interactions.
  • Widely used as an injectable anticoagulant.
  • Has the highest negative charge density of any known biological molecule.
  • Significantly inhibits interactions between exosomes and cells.
  • Intended for research purposes.
Catalog No. 50-003-85313 Supplier Medchemexpress LLC Supplier No. 250MGHY17567
Heparin is a highly sulfated glycosaminoglycan that is widely used as an injectable anticoagulant. It possesses the highest negative charge density of any known biological molecule and significantly inhibits exosome-cell interactions.
  • Widely used as an injectable anticoagulant.
  • Has the highest negative charge density of any known biological molecule.
  • Significantly inhibits interactions between exosomes and cells.
  • Intended for research purposes.